missing translation for 'onlineSavingsMsg'
Learn More

3,4-Difluorobenzenesulfonyl chloride, 97%

Catalog No. p-7047164
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAL1781603 1 g
AAL1781606 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAL1781603 Supplier Thermo Scientific Chemicals Supplier No. L1781603

CAS: 145758-05-0 | C6H3ClF2O2S | 212.595 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 145758-05-0
Molecular Formula C6H3ClF2O2S
Molecular Weight (g/mol) 212.595
MDL Number MFCD00153120
InChI Key FSGLUBQENACWCC-UHFFFAOYSA-N
Synonym 3,4-difluorobenzene-1-sulfonyl chloride, 3,4-difluorobenzenesulphonyl chloride, 3,4-difluoro-benzenesulfonyl chloride, benzenesulfonyl chloride, 3,4-difluoro, 3,4-difluorophenyl chlorosulfone, 3,4-difluorobenzenesulfonylchloride, 3,4-difluorobenzene sulfonyl chloride, 3,4-difluorobenzenesulphonylchloride, pubchem1958, acmc-1byq8
PubChem CID 2736913
IUPAC Name 3,4-difluorobenzenesulfonyl chloride
SMILES C1=CC(=C(C=C1S(=O)(=O)Cl)F)F

Specifications

Density 1.586
Boiling Point 105°C (15 mmHg)
Refractive Index 1.514
Quantity 1 g
UN Number UN3265
Beilstein 4802506
Sensitivity Moisture sensitive
Formula Weight 212.6
Percent Purity 97%
Chemical Name or Material 3,4-Difluorobenzenesulfonyl chloride
Hazard Category H314-H335
Hazard Statement GHS H Statement
H314-H318
Causes severe skin burns and eye damage.
Causes serious eye damage.
Precautionary Statement P260-P264b-P271-P280-P301+P330+P331-P303+P361+P353-P304+P340-P305+P351+P338-P310-P363-P501c
DOTInformation Transport Hazard Class: 8; Packing Group: II; Proper Shipping Name: CORROSIVE LIQUID, ACIDIC, ORGANIC, N.O.S.
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only