missing translation for 'onlineSavingsMsg'
Learn More
Learn More
3',4'-Difluoroacetophenone 97.0+%, TCI America™
$50.33 - $143.54
Chemical Identifiers
| CAS | 369-33-5 |
|---|---|
| Molecular Formula | C8H6F2O |
| Molecular Weight (g/mol) | 156.132 |
| MDL Number | MFCD00009891 |
| InChI Key | VWJSSJFLXRMYNV-UHFFFAOYSA-N |
| Synonym | 3',4'-difluoroacetophenone, 1-3,4-difluorophenyl ethanone, 3,4-difluoroacetophenone, 1-3,4-difluorophenyl ethan-1-one, ethanone, 1-3,4-difluorophenyl, 1-3,4-difluoro-phenyl-ethanone, 3,4,-difluoroacetophenone, 1-acetyl-3,4-difluorobenzene, pubchem3089, acmc-209ipo |
| PubChem CID | 123052 |
| IUPAC Name | 1-(3,4-difluorophenyl)ethanone |
| SMILES | CC(=O)C1=CC(=C(C=C1)F)F |
Chemical Identifiers
| 369-33-5 | |
| 156.132 | |
| VWJSSJFLXRMYNV-UHFFFAOYSA-N | |
| 123052 | |
| CC(=O)C1=CC(=C(C=C1)F)F |
| C8H6F2O | |
| MFCD00009891 | |
| 3',4'-difluoroacetophenone, 1-3,4-difluorophenyl ethanone, 3,4-difluoroacetophenone, 1-3,4-difluorophenyl ethan-1-one, ethanone, 1-3,4-difluorophenyl, 1-3,4-difluoro-phenyl-ethanone, 3,4,-difluoroacetophenone, 1-acetyl-3,4-difluorobenzene, pubchem3089, acmc-209ipo | |
| 1-(3,4-difluorophenyl)ethanone |
Specifications
| 369-33-5 | |
| 94°C | |
| MFCD00009891 | |
| 3',4'-difluoroacetophenone, 1-3,4-difluorophenyl ethanone, 3,4-difluoroacetophenone, 1-3,4-difluorophenyl ethan-1-one, ethanone, 1-3,4-difluorophenyl, 1-3,4-difluoro-phenyl-ethanone, 3,4,-difluoroacetophenone, 1-acetyl-3,4-difluorobenzene, pubchem3089, acmc-209ipo | |
| CC(=O)C1=CC(=C(C=C1)F)F | |
| 156.132 | |
| 156.13 | |
| Liquid |
| Yellow | |
| C8H6F2O | |
| 5 g | |
| VWJSSJFLXRMYNV-UHFFFAOYSA-N | |
| 1-(3,4-difluorophenyl)ethanone | |
| 123052 | |
| ≥97.0% (GC) | |
| 3′,4′-Difluoroacetophenone |
Safety and Handling
TSCA : No