missing translation for 'onlineSavingsMsg'
Learn More

3',4'-Difluoroacetophenone 97.0+%, TCI America™

Catalog No. D19065G
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
D19065G 5 g
D190625G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. D19065G Supplier TCI America Supplier No. D19065G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 369-33-5
Molecular Formula C8H6F2O
Molecular Weight (g/mol) 156.132
MDL Number MFCD00009891
InChI Key VWJSSJFLXRMYNV-UHFFFAOYSA-N
Synonym 3',4'-difluoroacetophenone, 1-3,4-difluorophenyl ethanone, 3,4-difluoroacetophenone, 1-3,4-difluorophenyl ethan-1-one, ethanone, 1-3,4-difluorophenyl, 1-3,4-difluoro-phenyl-ethanone, 3,4,-difluoroacetophenone, 1-acetyl-3,4-difluorobenzene, pubchem3089, acmc-209ipo
PubChem CID 123052
IUPAC Name 1-(3,4-difluorophenyl)ethanone
SMILES CC(=O)C1=CC(=C(C=C1)F)F

Specifications

Color Yellow
Boiling Point 94°C
Quantity 5 g
Formula Weight 156.13
Percent Purity ≥97.0% (GC)
Physical Form Liquid
Chemical Name or Material 3′,4′-Difluoroacetophenone