missing translation for 'onlineSavingsMsg'
Learn More

3',4'-Difluoroacetophenone, 98%

Catalog No. AAA1583222
Change view
Click to view available options
Quantity:
5 g
25 g
100 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAA1583222 100 g
AAA1583206 5 g
AAA1583214 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AAA1583222 Supplier Thermo Scientific Chemicals Supplier No. A1583222
Get Up to $1,800 in Chemicals with Purchase
View all promotions

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 369-33-5 | C8H6F2O | 156.132 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 369-33-5
Molecular Formula C8H6F2O
Molecular Weight (g/mol) 156.132
MDL Number MFCD00009891
InChI Key VWJSSJFLXRMYNV-UHFFFAOYSA-N
Synonym 3',4'-difluoroacetophenone, 1-3,4-difluorophenyl ethanone, 3,4-difluoroacetophenone, 1-3,4-difluorophenyl ethan-1-one, ethanone, 1-3,4-difluorophenyl, 1-3,4-difluoro-phenyl-ethanone, 3,4,-difluoroacetophenone, 1-acetyl-3,4-difluorobenzene, pubchem3089, acmc-209ipo
PubChem CID 123052
IUPAC Name 1-(3,4-difluorophenyl)ethanone
SMILES CC(=O)C1=CC(=C(C=C1)F)F

Specifications

Melting Point 19°C to 20°C
Density 1.246
Boiling Point 94°C to 95°C (13 mmHg)
Flash Point 75°C (167°F)
Odor Characteristic
Refractive Index 1.4916
Quantity 100 g
Beilstein 2206856
Formula Weight 156.13
Percent Purity 98%
Chemical Name or Material 3',4'-Difluoroacetophenone
Show More Show Less
Hazard Category H227-H315-H319-H335
Hazard Statement GHS H Statement
H315-H319-H335-H227
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Combustible liquid.
Precautionary Statement P210-P235-P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P370+P378q-P501c
EINECSNumber 206-717-7
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only