missing translation for 'onlineSavingsMsg'
Learn More

3,4-Difluoro-N-Fmoc-L-phenylalanine, 95%

Catalog No. p-7034374
Change view
Click to view available options
Quantity:
250 mg
1 g
5 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAH52057MD 250 mg
AAH5205703 1 g
AAH5205706 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AAH52057MD Supplier Thermo Scientific Chemicals Supplier No. H52057MD
Only null left

CAS: 198560-43-9 | C24H19F2NO4 | 423.416 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 198560-43-9
Molecular Formula C24H19F2NO4
Molecular Weight (g/mol) 423.416
MDL Number MFCD00672550
InChI Key IHSYIDJNVXPQRM-QFIPXVFZSA-N
Synonym fmoc-3,4-difluoro-l-phenylalanine, fmoc-phe 3,4-dif-oh, fmoc-l-3,4-difluorophe, fmoc-l-3,4-difluorophenylalanine, s-2-9h-fluoren-9-yl methoxy carbonyl amino-3-3,4-difluorophenyl propanoic acid, fmoc-phe 3,4-f2-oh, 3,4-difluoro-n-fmoc-l-phenylalanine, fmoc-d-3,4-difluorophe, 2s-3-3,4-difluorophenyl-2-9h-fluoren-9-ylmethoxy carbonyl amino propanoic acid
PubChem CID 12135342
IUPAC Name (2S)-3-(3,4-difluorophenyl)-2-(9H-fluoren-9-ylmethoxycarbonylamino)propanoic acid
SMILES C1=CC=C2C(=C1)C(C3=CC=CC=C32)COC(=O)NC(CC4=CC(=C(C=C4)F)F)C(=O)O

Specifications

Quantity 250 mg
Beilstein 8947230
Formula Weight 423.4
Percent Purity 95%
Chemical Name or Material 3,4-Difluoro-N-Fmoc-L-phenylalanine
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only