missing translation for 'onlineSavingsMsg'
Learn More

3,4-Difluoro-4'-(trans-4-propylcyclohexyl)biphenyl 98.0+%, TCI America™

Supplier:  TCI America D453525G

 View more versions of this product

Catalog No. D453525G

Only null left
Explore available promotions

Description

Description

Specifications

Specifications

3,4-Difluoro-4′-(trans-4-propylcyclohexyl)biphenyl
68°C
C21H24F2
25 g
VULXHDGYVHCLLN-UHFFFAOYSA-N
3,4-difluoro-4'-(4-propylcyclohexyl)-1,1'-biphenyl
612306
≥98.0% (GC)
85312-59-0
White
MFCD09838999
4'-trans-4-propylcyclohexyl-3,4-difluorobiphenyl, 3,4-difluoro-4'-trans-4-propylcyclohexyl-1,1'-biphenyl, 1,1'-biphenyl, 3,4-difluoro-4'-trans-4-propylcyclohexyl, 3,4-difluoro-4'-trans-4-propylcyclohexyl biphenyl, 1,2-difluoro-4-4-4-propylcyclohexyl phenyl benzene, 3,4-difluoro-4'-1s,4r-4-propylcyclohexyl-1,1'-biphenyl, 3,4-difluoro-4'-4-propylcyclohexyl-1,1'-biphenyl, 3,4-difluoro-4'-4-propylcyclohexyl biphenyl, 4-4-propylcyclohexyl-3',4'-difluorobiphenyl, trans-3,4-difluoro-4'-4-n-propylcyclohexyl biphenyl
CCCC1CCC(CC1)C1=CC=C(C=C1)C1=CC(F)=C(F)C=C1
314.42
314.42
Crystalline Powder

Chemical Identifiers

85312-59-0
314.42
VULXHDGYVHCLLN-UHFFFAOYSA-N
612306
CCCC1CCC(CC1)C1=CC=C(C=C1)C1=CC(F)=C(F)C=C1
C21H24F2
MFCD09838999
4'-trans-4-propylcyclohexyl-3,4-difluorobiphenyl, 3,4-difluoro-4'-trans-4-propylcyclohexyl-1,1'-biphenyl, 1,1'-biphenyl, 3,4-difluoro-4'-trans-4-propylcyclohexyl, 3,4-difluoro-4'-trans-4-propylcyclohexyl biphenyl, 1,2-difluoro-4-4-4-propylcyclohexyl phenyl benzene, 3,4-difluoro-4'-1s,4r-4-propylcyclohexyl-1,1'-biphenyl, 3,4-difluoro-4'-4-propylcyclohexyl-1,1'-biphenyl, 3,4-difluoro-4'-4-propylcyclohexyl biphenyl, 4-4-propylcyclohexyl-3',4'-difluorobiphenyl, trans-3,4-difluoro-4'-4-n-propylcyclohexyl biphenyl
3,4-difluoro-4'-(4-propylcyclohexyl)-1,1'-biphenyl
Product Suggestions

Product Suggestions

Videos
Safety and Handling

Safety and Handling

TSCA : No

SDS
Documents

Documents

Promotions

Promotions