missing translation for 'onlineSavingsMsg'
Learn More

3,4-Difluoro-4'-(trans-4-propylcyclohexyl)biphenyl 98.0+%, TCI America™

Catalog No. D45355G
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
D45355G 5 g
D453525G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. D45355G Supplier TCI America Supplier No. D45355G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 85312-59-0
Molecular Formula C21H24F2
Molecular Weight (g/mol) 314.42
MDL Number MFCD09838999
InChI Key VULXHDGYVHCLLN-UHFFFAOYSA-N
Synonym 4'-trans-4-propylcyclohexyl-3,4-difluorobiphenyl, 3,4-difluoro-4'-trans-4-propylcyclohexyl-1,1'-biphenyl, 1,1'-biphenyl, 3,4-difluoro-4'-trans-4-propylcyclohexyl, 3,4-difluoro-4'-trans-4-propylcyclohexyl biphenyl, 1,2-difluoro-4-4-4-propylcyclohexyl phenyl benzene, 3,4-difluoro-4'-1s,4r-4-propylcyclohexyl-1,1'-biphenyl, 3,4-difluoro-4'-4-propylcyclohexyl-1,1'-biphenyl, 3,4-difluoro-4'-4-propylcyclohexyl biphenyl, 4-4-propylcyclohexyl-3',4'-difluorobiphenyl, trans-3,4-difluoro-4'-4-n-propylcyclohexyl biphenyl
PubChem CID 612306
IUPAC Name 3,4-difluoro-4'-(4-propylcyclohexyl)-1,1'-biphenyl
SMILES CCCC1CCC(CC1)C1=CC=C(C=C1)C1=CC(F)=C(F)C=C1

Specifications

Melting Point 68°C
Color White
Quantity 5 g
Formula Weight 314.42
Percent Purity ≥98.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material 3,4-Difluoro-4′-(trans-4-propylcyclohexyl)biphenyl