missing translation for 'onlineSavingsMsg'
Learn More
Learn More
3,4-Difluoro-4'-(trans-4-pentylcyclohexyl)biphenyl 98.0+%, TCI America™
Supplier: TCI America D453425G
Specifications
| 3,4-Difluoro-4′-(trans-4-pentylcyclohexyl)biphenyl | |
| 58°C | |
| C23H28F2 | |
| 25 g | |
| NQLHGECGZRJQLF-UHFFFAOYSA-N | |
| 3,4-difluoro-4'-(4-pentylcyclohexyl)-1,1'-biphenyl | |
| 612307 | |
| ≥98.0% (GC) |
| 134412-17-2 | |
| White | |
| MFCD09839000 | |
| 3,4-difluoro-4'-trans-4-pentylcyclohexyl-1,1'-biphenyl, 3,4-difluoro-4'-4-pentylcyclohexyl biphenyl, 1,2-difluoro-4-4-4-pentylcyclohexyl phenyl benzene, 1,1'-biphenyl, 3,4-difluoro-4'-trans-4-pentylcyclohexyl, 1,1'-biphenyl,3,4-difluoro-4'-trans-4-pentylcyclohexyl, 3,4-difluoro-4'-1s,4r-4-pentylcyclohexyl-1,1'-biphenyl, acmc-20mvco, 3,4-difluoro-4/'-4-pentylcyclohexyl biphenyl, 3,4-difluoro-4'-trans-4-pentylcyclohexyl biphenyl, 1-3,4-difluorophenyl-4-4-pentylcyclohexyl benzene | |
| CCCCCC1CCC(CC1)C1=CC=C(C=C1)C1=CC(F)=C(F)C=C1 | |
| 342.47 | |
| 342.47 | |
| Crystalline Powder |
Chemical Identifiers
| 134412-17-2 | |
| 342.47 | |
| NQLHGECGZRJQLF-UHFFFAOYSA-N | |
| 612307 | |
| CCCCCC1CCC(CC1)C1=CC=C(C=C1)C1=CC(F)=C(F)C=C1 |
| C23H28F2 | |
| MFCD09839000 | |
| 3,4-difluoro-4'-trans-4-pentylcyclohexyl-1,1'-biphenyl, 3,4-difluoro-4'-4-pentylcyclohexyl biphenyl, 1,2-difluoro-4-4-4-pentylcyclohexyl phenyl benzene, 1,1'-biphenyl, 3,4-difluoro-4'-trans-4-pentylcyclohexyl, 1,1'-biphenyl,3,4-difluoro-4'-trans-4-pentylcyclohexyl, 3,4-difluoro-4'-1s,4r-4-pentylcyclohexyl-1,1'-biphenyl, acmc-20mvco, 3,4-difluoro-4/'-4-pentylcyclohexyl biphenyl, 3,4-difluoro-4'-trans-4-pentylcyclohexyl biphenyl, 1-3,4-difluorophenyl-4-4-pentylcyclohexyl benzene | |
| 3,4-difluoro-4'-(4-pentylcyclohexyl)-1,1'-biphenyl |
Safety and Handling
TSCA : No