missing translation for 'onlineSavingsMsg'
Learn More

3,4-Difluoro-4'-(trans-4-ethylcyclohexyl)biphenyl 98.0+%, TCI America™

Catalog No. p-7128507
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
D48595G 5 g
D485925G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. D48595G Supplier TCI America Supplier No. D48595G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 134412-18-3
Molecular Formula C20H22F2
Molecular Weight (g/mol) 300.39
MDL Number MFCD12828121
InChI Key TYZNCUASDJPXMP-UHFFFAOYSA-N
Synonym 3,4-difluoro-4'-4-ethylcyclohexyl biphenyl, 3,4-difluoro-4'-trans-4-ethylcyclohexyl biphenyl, 4'-4-ethylcyclohexyl-3,4-difluoro-1,1'-biphenyl, 3,4-difluoro-4/'-4-ethylcyclohexyl biphenyl, 1-3,4-difluorophenyl-4-4-ethylcyclohexyl benzene, 4-4-4-ethylcyclohexyl phenyl-1,2-difluorobenzene, 1,1'-biphenyl, 4'-4-ethylcyclohexyl-3,4-difluoro, 4'-4alpha-ethylcyclohexane-1beta-yl-3,4-difluorobiphenyl, 4-4-ethylcyclohexyl-3',4'-difluoro-1,1'-biphenyl #
PubChem CID 612308
IUPAC Name 4'-(4-ethylcyclohexyl)-3,4-difluoro-1,1'-biphenyl
SMILES CCC1CCC(CC1)C1=CC=C(C=C1)C1=CC(F)=C(F)C=C1

Specifications

Melting Point 69°C
Color White
Quantity 5 g
Formula Weight 300.39
Percent Purity ≥98.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material 3,4-Difluoro-4′-(trans-4-ethylcyclohexyl)biphenyl