missing translation for 'onlineSavingsMsg'
Learn More

3,4-Diethyl-2-ethoxycarbonyl-5-methylpyrrole 96.0+%, TCI America™

Catalog No. p-7127174
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
D22171G 1 g
D22175G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. D22171G Supplier TCI America Supplier No. D22171G
Only null left

Chemical Identifiers

CAS 16200-50-3
Molecular Formula C12H19NO2
Molecular Weight (g/mol) 209.289
MDL Number MFCD00075089
InChI Key YXAABSBFWADBRO-UHFFFAOYSA-N
Synonym 3,4-Diethyl-5-methyl-2-pyrrolecarboxylic Acid Ethyl Ester, Ethyl 3,4-Diethyl-5-methyl-2-pyrrolecarboxylate
PubChem CID 590710
IUPAC Name ethyl 3,4-diethyl-5-methyl-1H-pyrrole-2-carboxylate
SMILES CCC1=C(NC(=C1CC)C(=O)OCC)C

Specifications

Melting Point 76°C
Color White-Yellow
Quantity 1 g
Formula Weight 209.29
Percent Purity ≥96.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material 3,4-Diethyl-2-ethoxycarbonyl-5-methylpyrrole