missing translation for 'onlineSavingsMsg'
Learn More

3,4-Dichlorotoluene 98.0+%, TCI America™

Catalog No. D0426500G
Change view
Click to view available options
Quantity:
25 g
500 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
D0426500G 500 g
D042625G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. D0426500G Supplier TCI America Supplier No. D0426500G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 95-75-0
Molecular Formula C7H6Cl2
Molecular Weight (g/mol) 161.025
MDL Number MFCD00000556
InChI Key WYUIWKFIFOJVKW-UHFFFAOYSA-N
Synonym 3,4-dichlorotoluene, benzene, 1,2-dichloro-4-methyl, toluene, 3,4-dichloro, 3,4-dichloro-toluene, ksc486q9r, chembl47525, benzene,2-dichloro-4-methyl, 1,2-dichloro-4-methyl-benzene, #
PubChem CID 7256
IUPAC Name 1,2-dichloro-4-methylbenzene
SMILES CC1=CC(=C(C=C1)Cl)Cl

Specifications

Color Colorless
Boiling Point 205°C
Quantity 500 g
UN Number 3082
Formula Weight 161.03
Percent Purity ≥98.0% (GC)
Physical Form Liquid
Chemical Name or Material 3,4-Dichlorotoluene
EINECSNumber (3)-0078
TSCA Yes