missing translation for 'onlineSavingsMsg'
Learn More

3,4-Dichloropyridine, 98%

Catalog No. AAH6374906
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAH6374906 5 g
AAH6374903 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAH6374906 Supplier Thermo Scientific Chemicals Supplier No. H6374906
Get Up to $1,800 in Chemicals with Purchase
View all promotions

CAS: 55934-00-4 | C5H3Cl2N | 147.99 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 55934-00-4
Molecular Formula C5H3Cl2N
Molecular Weight (g/mol) 147.99
MDL Number MFCD01861989
InChI Key ZMPYQKNNUHPTLT-UHFFFAOYSA-N
Synonym pyridine, 3,4-dichloro, zlchem 375, 3,4,dichloropyridine, pubchem6613, acmc-209lq5, ksc493e3l
PubChem CID 2736081
IUPAC Name 3,4-dichloropyridine
SMILES ClC1=C(Cl)C=NC=C1

Specifications

Melting Point 22°C to 24°C
Color Colorless
Boiling Point 182°C to 183°C
Quantity 5 g
UN Number UN2810
Formula Weight 147.99
Percent Purity 98%
Physical Form Liquid
Chemical Name or Material 3,4-Dichloropyridine
Hazard Category H302+H312-H315-H319-H335
Hazard Statement GHS H Statement
H302-H312-H315-H319-H335
Harmful if swallowed.
Harmful in contact with skin.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P261-P264b-P270-P271-P280-P301+P312-P302+P352-P304+P340-P305+P351+P338-P312-P330-P332+P313-P362-P501c
DOTInformation Transport Hazard Class: 6.1; Packaging Group: III; Proper Shipping Name: TOXIC, LIQUIDS, ORGANIC, N.O.S.
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only