missing translation for 'onlineSavingsMsg'
Learn More

3,4-Dichlorophenylacetic Acid 98.0+%, TCI America™

Catalog No. p-7126944
Change view
missing translation for 'orderingAttributeHoverText'
Quantity:
25 g
500 g
Catalog No. Quantity
D181325G 25 g
D1813500G 500 g
2 options

Catalog No. D181325G

Supplier: TCI America D181325G

Only null left

Chemical Identifiers

CAS 5807-30-7
Molecular Formula C8H6Cl2O2
Molecular Weight (g/mol) 205.034
MDL Number MFCD00004333
InChI Key ZOUPGSMSNQLUNW-UHFFFAOYSA-N
Synonym 3,4-dichlorophenylacetic acid, 2-3,4-dichlorophenyl acetic acid, 3,4-dichlorophenyl acetic acid, 3,4-dichlorobenzene acetic acid, benzeneacetic acid, 3,4-dichloro, pubchem20195, acmc-209m2g, 3,4dichlorophenylacetic acid, 3,4-dichlorophenylaceticacid
PubChem CID 79874
IUPAC Name 2-(3,4-dichlorophenyl)acetic acid
SMILES C1=CC(=C(C=C1CC(=O)O)Cl)Cl

Specifications

Melting Point 90°C
Color White
Quantity 25 g
Formula Weight 205.03
Percent Purity ≥98.0% (HPLC,T)
Physical Form Crystalline Powder
Chemical Name or Material 3,4-Dichlorophenylacetic Acid
EINECSNumber (3)-3805
TSCA No