missing translation for 'onlineSavingsMsg'
Learn More

3,4-Dichlorophenyl Isocyanate 98.0+%, TCI America™

$130.92 - $130.92

Chemical Identifiers

CAS 102-36-3
Molecular Formula C7H3Cl2NO
Molecular Weight (g/mol) 188.007
MDL Number MFCD00002017
InChI Key MFUVCHZWGSJKEQ-UHFFFAOYSA-N
Synonym 3,4-dichlorophenyl isocyanate, benzene, 1,2-dichloro-4-isocyanato, 3,4-dichlorfenylisokyanat, isocyanic acid 3,4-dichlorophenyl ester, 3,4-dichlorophenylisocyanate, unii-ozw0b7f1du, 3,4-dichlorphenylisocyanate, isocyanic acid, 3,4-dichlorophenyl ester, 3,4-dichlorfenylisokyanat czech, ozw0b7f1du
PubChem CID 7607
IUPAC Name 1,2-dichloro-4-isocyanatobenzene
SMILES C1=CC(=C(C=C1N=C=O)Cl)Cl
View More Specs

Products 1
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
I009225G
View Documents
TCI America
I009225G
25 g
Each for $130.92
Only null left
 
Description

Description

Specifications

Chemical Identifiers

102-36-3
188.007
MFUVCHZWGSJKEQ-UHFFFAOYSA-N
7607
C1=CC(=C(C=C1N=C=O)Cl)Cl
C7H3Cl2NO
MFCD00002017
3,4-dichlorophenyl isocyanate, benzene, 1,2-dichloro-4-isocyanato, 3,4-dichlorfenylisokyanat, isocyanic acid 3,4-dichlorophenyl ester, 3,4-dichlorophenylisocyanate, unii-ozw0b7f1du, 3,4-dichlorphenylisocyanate, isocyanic acid, 3,4-dichlorophenyl ester, 3,4-dichlorfenylisokyanat czech, ozw0b7f1du
1,2-dichloro-4-isocyanatobenzene

Specifications

102-36-3
White-Yellow
C7H3Cl2NO
25 g
3,4-dichlorophenyl isocyanate, benzene, 1,2-dichloro-4-isocyanato, 3,4-dichlorfenylisokyanat, isocyanic acid 3,4-dichlorophenyl ester, 3,4-dichlorophenylisocyanate, unii-ozw0b7f1du, 3,4-dichlorphenylisocyanate, isocyanic acid, 3,4-dichlorophenyl ester, 3,4-dichlorfenylisokyanat czech, ozw0b7f1du
C1=CC(=C(C=C1N=C=O)Cl)Cl
188.007
188.01
Crystalline Lumps
40°C
120°C
MFCD00002017
2250
MFUVCHZWGSJKEQ-UHFFFAOYSA-N
1,2-dichloro-4-isocyanatobenzene
7607
≥98.0% (GC,T)
3,4-Dichlorophenyl Isocyanate
Videos
Safety and Handling

Safety and Handling

EINECSNumber : (3)-2489

RTECSNumber : NQ8760000

TSCA : Yes

Recommended Storage : Refrigerator

SDS
Documents

Documents