missing translation for 'onlineSavingsMsg'
Learn More

3,4-Dichlorophenethyl alcohol, 97%

Catalog No. p-106529
Change view
Click to view available options
Quantity:
1 g
5 g
Packaging:
Glass Bottle
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
AC350770010 1 g Glass Bottle
AC350770050 5 g Glass Bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AC350770010 Supplier Thermo Scientific Chemicals Supplier No. 350770010

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 35364-79-5 | C8H8Cl2O | 191.051 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 35364-79-5
Molecular Formula C8H8Cl2O
Molecular Weight (g/mol) 191.051
MDL Number MFCD00800673
InChI Key GITOMJDYNUMCOV-UHFFFAOYSA-N
Synonym 2-3,4-dichlorophenyl ethanol, 3,4-dichlorophenethyl alcohol, 3,4-dichlorophenethylalcohol, 2-3,4-dichlorophenyl ethan-1-ol, benzeneethanol, 3,4-dichloro, 2-3,4-dichlorophenyl-ethanol, 3,4-dichlorobenzeneethanol, benzeneethanol,3,4-dichloro, acmc-1adlk, 2-3,4-dichloro-phenyl-ethanol
PubChem CID 244558
IUPAC Name 2-(3,4-dichlorophenyl)ethanol
SMILES C1=CC(=C(C=C1CCO)Cl)Cl

Specifications

CAS Min % 96.0
CAS Max % 100.0
Color Colorless
Infrared Spectrum Authentic
Assay Percent Range 97%
Packaging Glass Bottle
Refractive Index 1.5620 to 1.5680 (20°C, 589nm)
Quantity 1 g
Beilstein 06, IV, 3081
Formula Weight 191.06
Percent Purity 97%
Physical Form Liquid
Chemical Name or Material 3, 4-Dichlorophenethyl alcohol, 97%
Show More Show Less
Hazard Category GHS Signal Word: Warning
Hazard Statement GHS H Statement:
Harmful if swallowed.
Causes skin irritation.
Causes serious eye irritation.
Harmful in contact with skin.
Harmful if inhaled.
May cause respiratory irritation.
Precautionary Statement GHS P Statement:
Wear protective gloves/protective clothing/eye protection/face protection.
IF ON SKIN: Wash with plenty of soap and water.
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove victim to fresh air and keep at rest in a position comfortable for breathing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
TSCA TSCA

RUO – Research Use Only