missing translation for 'onlineSavingsMsg'
Learn More

3,4-Dichlorophenacyl Bromide 98.0+%, TCI America™

Catalog No. p-7119794
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
B28375G 5 g
B283725G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. B28375G Supplier TCI America Supplier No. B28375G

Chemical Identifiers

CAS 2632-10-2
Molecular Formula C8H5BrCl2O
Molecular Weight (g/mol) 267.93
InChI Key PAKFHEFMTRCFAU-UHFFFAOYSA-N
Synonym 2-bromo-1-3,4-dichlorophenyl ethanone, 3,4-dichlorophenacyl bromide, 2-bromo-3',4'-dichloroacetophenone, 2-bromo-1-3,4-dichlorophenyl ethan-1-one, 3,4-dichlorophenacylbromide, ethanone, 2-bromo-1-3,4-dichlorophenyl, 1-3,4-dichlorophenyl-2-bromoethan-1-one, pubchem3262, acmc-1coei, 3.4-dichlorophenacylbromide
PubChem CID 244751
IUPAC Name 2-bromo-1-(3,4-dichlorophenyl)ethanone
SMILES C1=CC(=C(C=C1C(=O)CBr)Cl)Cl

Specifications

Melting Point 58°C
Color White-Yellow
Quantity 5 g
UN Number 3261
Formula Weight 267.93
Percent Purity ≥98.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material 3,4-Dichlorophenacyl Bromide