missing translation for 'onlineSavingsMsg'
Learn More

3,4-Dichlorocinnamic acid, 97%

Catalog No. p-7028158
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAB2530806 5 g
AAB2530814 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAB2530806 Supplier Thermo Scientific Chemicals Supplier No. B2530806
Only null left

CAS: 1202-39-7 | C9H6Cl2O2 | 217.045 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 1202-39-7
Molecular Formula C9H6Cl2O2
Molecular Weight (g/mol) 217.045
MDL Number MFCD00004385
InChI Key RRLUFPHCTSFKNR-DUXPYHPUSA-N
Synonym 3,4-dichlorocinnamic acid, e-3-3,4-dichlorophenyl acrylic acid, 3',4'-dichlorocinnamic acid, 2-propenoic acid, 3-3,4-dichlorophenyl, cinnamic acid, 3,4-dichloro, 3-3,4-dichlorophenyl acrylic acid, 2e-3-3,4-dichlorophenyl prop-2-enoic acid, 3-3,4-dichlorophenyl-2-propenoic acid, e-3-3,4-dichlorophenyl prop-2-enoic acid, 2e-3-3,4-dichlorophenyl acrylic acid
PubChem CID 688027
IUPAC Name (E)-3-(3,4-dichlorophenyl)prop-2-enoic acid
SMILES C1=CC(=C(C=C1C=CC(=O)O)Cl)Cl

Specifications

Melting Point 218°C to 220°C
Quantity 5 g
Beilstein 1872129
Formula Weight 217.05
Percent Purity 97%
Chemical Name or Material 3,4-Dichlorocinnamic acid
Hazard Category H315-H319-H335
Hazard Statement GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
EINECSNumber 214-866-4
RTECSNumber UD3333900
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only