missing translation for 'onlineSavingsMsg'
Learn More

3,4-Dichlorobenzyl mercaptan, 97%

Catalog No. p-7043873
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAL0322603 1 g
AAL0322606 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAL0322603 Supplier Thermo Scientific Chemicals Supplier No. L0322603

CAS: 36480-40-7 | C7H6Cl2S | 193.085 g/mol

Reatant in the synthesis of substituted pyridazines. 3,4-Dichlorobenzyl mercaptan helps to improve the semiconductor morphologies.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Reatant in the synthesis of substituted pyridazines. 3,4-Dichlorobenzyl mercaptan helps to improve the semiconductor morphologies.

Solubility
Difficult to mix in water.

Notes
Store at room temperature. Incompatible with oxidizing agents. Ensure good ventilation.

Chemical Identifiers

CAS 36480-40-7
Molecular Formula C7H6Cl2S
Molecular Weight (g/mol) 193.085
MDL Number MFCD00039651
InChI Key CEGBRSQPRQXALB-UHFFFAOYSA-N
Synonym 3,4-dichlorobenzyl mercaptan, 3,4-dichlorobenzenemethanethiol, 3,4-dichlorophenyl methanethiol, 3,4-dichlorotoluene-alpha-thiol, benzenemethanethiol, 3,4-dichloro, acmc-1aex5, d0z0bb, 3,4-dichlorobenzylmercaptan, 3,4-dichloro-alpha-toluenethiol, 3,4-dichloro-phenyl-methanethiol
PubChem CID 118986
IUPAC Name (3,4-dichlorophenyl)methanethiol
SMILES C1=CC(=C(C=C1CS)Cl)Cl

Specifications

Density 1.362
Boiling Point 152°C to 153°C (12 mmHg)
Flash Point >110°C (230°F)
Refractive Index 1.607
Quantity 1 g
UN Number UN2810
Beilstein 3237473
Sensitivity Air Sensitive
Solubility Information Difficult to mix in water.
Formula Weight 193.1
Percent Purity 97%
Chemical Name or Material 3,4-Dichlorobenzyl mercaptan
Show More Show Less
Hazard Category H302-H315-H319-H331-H335
Hazard Statement GHS H Statement
H331-H302-H315-H319-H335
Toxic if inhaled.
Harmful if swallowed.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P261-P264b-P270-P271-P280-P301+P312-P302+P352-P304+P340-P305+P351+P338-P311-P330-P332+P313-P362-P501c
DOTInformation Transport Hazard Class: 6.1; Packing Group: III; Proper Shipping Name: TOXIC, LIQUIDS, ORGANIC, N.O.S.
EINECSNumber 253-054-4
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only