missing translation for 'onlineSavingsMsg'
Learn More

3,4-Dichlorobenzotrichloride 98.0+%, TCI America™

Catalog No. D248125G
Change view
Click to view available options
Quantity:
25 g
500 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
D248125G 25 g
D2481500G 500 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. D248125G Supplier TCI America Supplier No. D248125G
Only null left

Chemical Identifiers

CAS 13014-24-9
Molecular Formula C7H3Cl5
Molecular Weight (g/mol) 264.35
MDL Number MFCD00018820
InChI Key ATYLRBXENHNROH-UHFFFAOYSA-N
Synonym alpha,alpha,alpha,3,4-Pentachlorotoluene
PubChem CID 25608
IUPAC Name 1,2-dichloro-4-(trichloromethyl)benzene
SMILES ClC1=CC=C(C=C1Cl)C(Cl)(Cl)Cl

Specifications

Color White
Quantity 25 g
UN Number 1759
Formula Weight 264.35
Percent Purity ≥98.0% (GC)
Physical Form Crystalline Lumps
Chemical Name or Material 3,4-Dichlorobenzotrichloride
EINECSNumber (3)-0093
RTECSNumber CZ5475000
TSCA Yes