missing translation for 'onlineSavingsMsg'
Learn More
Learn More
3,4-Dichloroaniline, 98%
Description
Ce produit de marque Thermo Scientific Chemicals faisait à l’origine partie du portefeuille Acros Organics. Certains documents et informations d’étiquetage peuvent faire référence à l’ancienne marque. Le code produit/article (SKU) original d’Acros Organics n’a pas changé dans le cadre de la transition vers la marque Thermo Scientific Chemicals.
Spécifications
Spécifications
| Nom ou substance chimique | 3, 4-Dichloroaniline |
| Nom Remarque | 98% |
| CAS % minimum | 97.5 |
| CAS % maximal | 100.0 |
| Point de fusion | 69,0 °C à 73,0 °C |
| Couleur | Beige à brun |
| Point d’ébullition | 272,0 °C |
| Point d’éclair | 166 °C |
| Spectre infrarouge | Authentique |
| Fourchette de pourcentages d’essai | 98% |
| Afficher plus de résultats |
Sécurité et manipulation
| Hazard Category | GHS Signal Word: Danger |
| Hazard Statement | GHS H Statement: Toxic if swallowed. May cause an allergic skin reaction. Toxic in contact with skin. Causes serious eye damage. Toxic if inhaled. Very toxic to aquatic life with long lasting effects. |
| Precautionary Statement | GHS P Statement: IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician. IF INHALED: Remove victim to fresh air and keep at rest in a position comfortable for breathing. IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing. IF ON SKIN: Gently wash with plenty of soap and water. Call a POISON CENTER or doctor/physician if you feel unwell. Wear protective gloves/protective clothing/eye protection/face protection. WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification. |
| EINECSNumber | 202-448-4 |
| RTECSNumber | BX2625000 |
| TSCA | TSCA |
| Recommended Storage | Le produit s’assombrit lors du stockage |
RUO – Usage réservé à la recherche