missing translation for 'onlineSavingsMsg'
Learn More

3,4-Dichloro-1,2,5-thiadiazole 98.0+%, TCI America™

Catalog No. D354725G
Change view
Click to view available options
Quantity:
25 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
D354725G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. D354725G Supplier TCI America Supplier No. D354725G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 5728-20-1
Molecular Formula C2Cl2N2S
Molecular Weight (g/mol) 155.00
MDL Number MFCD00010072
InChI Key YNZQOVYRCBAMEU-UHFFFAOYSA-N
Synonym 1,2,5-thiadiazole, 3,4-dichloro, dichloro-1,2,5-thiadiazole, 3,4-dichloro-1,2,5 thiadiazole, pubchem20790, acmc-1al9l, ksc272e3d, 4,5-dichloro-2,1,3-thiadiazole, 3,4-bis chloranyl-1,2,5-thiadiazole, 3,4-dichloro-1,2,5-thiadiazole
PubChem CID 79804
IUPAC Name dichloro-1,2,5-thiadiazole
SMILES ClC1=NSN=C1Cl

Specifications

Color Yellow
Boiling Point 158°C
Quantity 25 g
Formula Weight 155.00
Percent Purity ≥98.0% (GC)
Physical Form Liquid
Chemical Name or Material 3,4-Dichloro-1,2,5-thiadiazole