missing translation for 'onlineSavingsMsg'
Learn More
Learn More
3,4-Dibenzyloxybenzaldehyde, 99%, Thermo Scientific™
Chemical Identifiers
| CAS | 5447-02-9 |
|---|---|
| Molecular Formula | C21H18O3 |
| Molecular Weight (g/mol) | 318.36 |
| MDL Number | MFCD00004776 |
| InChI Key | XDDLXZHBWVFPRG-UHFFFAOYSA-N |
| Synonym | 3,4-dibenzyloxybenzaldehyde, 3,4-bis benzyloxy benzaldehyde, 3,4-dibenzyloxy benzaldehyde, 3,4-bis phenylmethoxy benzaldehyde, xddlxzhbwvfprg-uhfffaoysa-n, 3.4-dibenzyloxybenzaldehyde, 3,4-dibenzyloxy-benzaldehyde, 3,4-dibenzyl-oxybenzaldehyde, benzaldehyde, 3,4-dibenzyloxy, xddlxzhbwvfprg-uhfffaoysa |
| PubChem CID | 79526 |
| Catalog Number | Mfr. No. | Quantity | Packaging | Price | Quantity | ||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Quantity | Packaging | Price | Quantity | ||||
|
AC112640010
|
Thermo Scientific Chemicals
112640010 |
1g | Glass bottle |
N/A
|
N/A | ||||
| Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance. | |||||||||
|
AC112640050
|
Thermo Scientific Chemicals
112640050 |
5g | Glass bottle |
N/A
|
N/A | ||||
| Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance. | |||||||||
|
AC112640250
|
Thermo Scientific Chemicals
112640250 |
25g | Glass bottle |
N/A
|
N/A | ||||
| Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance. | |||||||||
Description
This Thermo Scientific brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.
Chemical Identifiers
| 5447-02-9 | |
| 318.36 | |
| XDDLXZHBWVFPRG-UHFFFAOYSA-N | |
| 79526 |
| C21H18O3 | |
| MFCD00004776 | |
| 3,4-dibenzyloxybenzaldehyde, 3,4-bis benzyloxy benzaldehyde, 3,4-dibenzyloxy benzaldehyde, 3,4-bis phenylmethoxy benzaldehyde, xddlxzhbwvfprg-uhfffaoysa-n, 3.4-dibenzyloxybenzaldehyde, 3,4-dibenzyloxy-benzaldehyde, 3,4-dibenzyl-oxybenzaldehyde, benzaldehyde, 3,4-dibenzyloxy, xddlxzhbwvfprg-uhfffaoysa |
Specifications
| 5447-02-9 | |
| 100.0 | |
| 98.5% min. (GC) | |
| C21H18O3 | |
| MFCD00004776 | |
| 3,4-dibenzyloxybenzaldehyde, 3,4-bis benzyloxy benzaldehyde, 3,4-dibenzyloxy benzaldehyde, 3,4-bis phenylmethoxy benzaldehyde, xddlxzhbwvfprg-uhfffaoysa-n, 3.4-dibenzyloxybenzaldehyde, 3,4-dibenzyloxy-benzaldehyde, 3,4-dibenzyl-oxybenzaldehyde, benzaldehyde, 3,4-dibenzyloxy, xddlxzhbwvfprg-uhfffaoysa | |
| 318.36 | |
| 318.36 | |
| 3, 4-Dibenzyloxybenzaldehyde |
| 98.5 | |
| Authentic | |
| Glass bottle | |
| (C6H5CH2O)2C6H3CHO | |
| 1g | |
| XDDLXZHBWVFPRG-UHFFFAOYSA-N | |
| 79526 | |
| 99% |
Safety and Handling
EINECSNumber : 226-662-2
RUO â Research Use Only