missing translation for 'onlineSavingsMsg'
Learn More
Learn More
3,4-Diaminopyridine 99.0+%, TCI America™
$65.24 - $192.00
Chemical Identifiers
| CAS | 54-96-6 |
|---|---|
| Molecular Formula | C5H7N3 |
| Molecular Weight (g/mol) | 109.132 |
| MDL Number | MFCD00006401 |
| InChI Key | OYTKINVCDFNREN-UHFFFAOYSA-N |
| Synonym | 3,4-diaminopyridine, amifampridine, 3,4-pyridinediamine, firdapse, diamino-3,4 pyridine, 4,5-diaminopyridine, 3,4-dap, amifampridin, zenas, pyridine, 3,4-diamino |
| PubChem CID | 5918 |
| IUPAC Name | pyridine-3,4-diamine |
| SMILES | C1=CN=CC(=C1N)N |
Chemical Identifiers
| 54-96-6 | |
| 109.132 | |
| OYTKINVCDFNREN-UHFFFAOYSA-N | |
| 5918 | |
| C1=CN=CC(=C1N)N |
| C5H7N3 | |
| MFCD00006401 | |
| 3,4-diaminopyridine, amifampridine, 3,4-pyridinediamine, firdapse, diamino-3,4 pyridine, 4,5-diaminopyridine, 3,4-dap, amifampridin, zenas, pyridine, 3,4-diamino | |
| pyridine-3,4-diamine |
Specifications
| 54-96-6 | |
| C5H7N3 | |
| 1 g | |
| 3,4-diaminopyridine, amifampridine, 3,4-pyridinediamine, firdapse, diamino-3,4 pyridine, 4,5-diaminopyridine, 3,4-dap, amifampridin, zenas, pyridine, 3,4-diamino | |
| C1=CN=CC(=C1N)N | |
| 109.132 | |
| 109.13 | |
| Crystalline Powder |
| 220°C | |
| MFCD00006401 | |
| 2811 | |
| OYTKINVCDFNREN-UHFFFAOYSA-N | |
| pyridine-3,4-diamine | |
| 5918 | |
| ≥99.0% (GC,T) | |
| 3,4-Diaminopyridine |
Safety and Handling
RTECSNumber : US7600000
TSCA : No