missing translation for 'onlineSavingsMsg'
Learn More

3,4-Diaminopyridine 99.0+%, TCI America™

Catalog No. D11495G
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
D11495G 5 g
D11491G 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. D11495G Supplier TCI America Supplier No. D11495G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 54-96-6 | C5H7N3 | 109.132 g/mol

Chemical Identifiers

CAS 54-96-6
Molecular Formula C5H7N3
Molecular Weight (g/mol) 109.132
MDL Number MFCD00006401
InChI Key OYTKINVCDFNREN-UHFFFAOYSA-N
Synonym 3,4-diaminopyridine, amifampridine, 3,4-pyridinediamine, firdapse, diamino-3,4 pyridine, 4,5-diaminopyridine, 3,4-dap, amifampridin, zenas, pyridine, 3,4-diamino
PubChem CID 5918
IUPAC Name pyridine-3,4-diamine
SMILES C1=CN=CC(=C1N)N

Specifications

Melting Point 220°C
Quantity 5 g
UN Number 2811
Formula Weight 109.13
Percent Purity ≥99.0% (GC,T)
Grade Not Available
Physical Form Crystalline Powder
Chemical Name or Material 3,4-Diaminopyridine
RTECSNumber US7600000
TSCA No