missing translation for 'onlineSavingsMsg'
Learn More

3,4-Diaminopyridine, 98%

Catalog No. AC112390250
Change view
Click to view available options
Quantity:
5 g
25 g
Packaging:
Glass bottle
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
AC112390250 25 g Glass bottle
AC112390050 5 g Glass bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AC112390250 Supplier Thermo Scientific Chemicals Supplier No. 112390250
Get Up to $1,800 in Chemicals with Purchase
View all promotions

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 54-96-6 | C5H7N3 | 109.13 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 54-96-6
Molecular Formula C5H7N3
Molecular Weight (g/mol) 109.13
MDL Number MFCD00006401
InChI Key OYTKINVCDFNREN-UHFFFAOYSA-N
Synonym 3,4-diaminopyridine, amifampridine, 3,4-pyridinediamine, firdapse, diamino-3,4 pyridine, 4,5-diaminopyridine, 3,4-dap, amifampridin, zenas, pyridine, 3,4-diamino
PubChem CID 5918
IUPAC Name pyridine-3,4-diamine
SMILES C1=CN=CC(=C1N)N

Specifications

CAS Min % 97.5
CAS Max % 100.0
Melting Point 216°C to 221°C
Color Brown to Brown-Gray
Flash Point 240°C
Infrared Spectrum Authentic
Assay Percent Range 97.5% min. (GC)
Packaging Glass bottle
Linear Formula (C5H3N)(NH2)2
Quantity 25 g
Beilstein 22, II, 395
Merck Index 15, 398
Solubility Information Solubility in water: 24g/L (20°C)
Formula Weight 109.13
Percent Purity 98%
Physical Form Crystalline Powder
Chemical Name or Material 3, 4-Diaminopyridine
Show More Show Less
Hazard Category GHS Signal Word: Danger
Hazard Statement GHS H Statement
Fatal if swallowed.
Toxic in contact with skin.
Fatal if inhaled.
Causes skin irritation.
Causes serious eye damage.
May cause respiratory irritation.
Precautionary Statement GHS P Statement
IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician.
Wear protective gloves/protective clothing/eye protection/face protection.
IF SWALLOWED: rinse mouth.
Do NOT induce vomiting.
Wear eye protection
EINECSNumber 200-220-9
Recommended Storage Product darkens on storage

RUO – Research Use Only