missing translation for 'onlineSavingsMsg'
Learn More

3,4-Diaminofurazan 98.0+%, TCI America™

$182.68 - $654.29

Chemical Identifiers

CAS 17220-38-1
Molecular Formula C2H4N4O
Molecular Weight (g/mol) 100.081
MDL Number MFCD00138084
InChI Key JHJVSUCUNFXIHN-UHFFFAOYSA-N
Synonym 3,4-diaminofurazan, furazandiamine, 3,4-diamino-1,2,5-oxadiazole, diaminofurazan, 4,5-diaminofurazan, 3,4-diaminofurazane, furazan-3,4-diamine, pubchem21086, #
PubChem CID 537677
IUPAC Name 1,2,5-oxadiazole-3,4-diamine
SMILES C1(=NON=C1N)N
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
D40301G
View Documents
TCI America
D40301G
1 g N/A
Only null left
 
D40305G
View Documents
TCI America
D40305G
5 g N/A
Only null left
 
Description

Description

Specifications

Chemical Identifiers

17220-38-1
100.081
JHJVSUCUNFXIHN-UHFFFAOYSA-N
537677
C1(=NON=C1N)N
C2H4N4O
MFCD00138084
3,4-diaminofurazan, furazandiamine, 3,4-diamino-1,2,5-oxadiazole, diaminofurazan, 4,5-diaminofurazan, 3,4-diaminofurazane, furazan-3,4-diamine, pubchem21086, #
1,2,5-oxadiazole-3,4-diamine

Specifications

17220-38-1
White-Yellow
MFCD00138084
3,4-diaminofurazan, furazandiamine, 3,4-diamino-1,2,5-oxadiazole, diaminofurazan, 4,5-diaminofurazan, 3,4-diaminofurazane, furazan-3,4-diamine, pubchem21086, #
C1(=NON=C1N)N
100.081
100.08
Crystalline Powder
182°C
C2H4N4O
1 g
JHJVSUCUNFXIHN-UHFFFAOYSA-N
1,2,5-oxadiazole-3,4-diamine
537677
≥98.0% (GC)
3,4-Diaminofurazan
Videos
Safety and Handling

Safety and Handling

TSCA : No

SDS
missing translation for 'documents'

missing translation for 'documents'