missing translation for 'onlineSavingsMsg'
Learn More

3,4-Diaminofurazan 98.0+%, TCI America™

Catalog No. D40301G
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
D40301G 1 g
D40305G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. D40301G Supplier TCI America Supplier No. D40301G

Chemical Identifiers

CAS 17220-38-1
Molecular Formula C2H4N4O
Molecular Weight (g/mol) 100.081
MDL Number MFCD00138084
InChI Key JHJVSUCUNFXIHN-UHFFFAOYSA-N
Synonym 3,4-diaminofurazan, furazandiamine, 3,4-diamino-1,2,5-oxadiazole, diaminofurazan, 4,5-diaminofurazan, 3,4-diaminofurazane, furazan-3,4-diamine, pubchem21086, #
PubChem CID 537677
IUPAC Name 1,2,5-oxadiazole-3,4-diamine
SMILES C1(=NON=C1N)N

Specifications

Melting Point 182°C
Color White-Yellow
Quantity 1 g
Formula Weight 100.08
Percent Purity ≥98.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material 3,4-Diaminofurazan