missing translation for 'onlineSavingsMsg'
Learn More

3,4-Di-O-acetyl-6-deoxy-L-glucal 95.0+%, TCI America™

Catalog No. D27525G
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
D27525G 5 g
D27521G 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. D27525G Supplier TCI America Supplier No. D27525G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 34819-86-8
Molecular Formula C10H14O5
Molecular Weight (g/mol) 214.217
MDL Number MFCD00074970
InChI Key NDEGMKQAZZBNBB-JUWDTYFHSA-N
Synonym 3,4-di-o-acetyl-6-deoxy-l-glucal, 3,4-di-o-acetyl-l-rhamnal, 2s,3s,4s-2-methyl-3,4-dihydro-2h-pyran-3,4-diyl diacetate, 3,4-di-o-acetyl-1,5-anhydro-2,6-dideoxy-l-arabino-hex-1-enitol, di-o-acetyl-l-rhamnal, 2s,3s,4s-4-acetyloxy-2-methyl-3,4-dihydro-2h-pyran-3-yl acetate, diacetyl-l-rhamnal, l-rhamnal diacetate, 6-deoxy-l-glucal diacetate, l-arabino-hex-1-enitol, 1,5-anhydro-2,6-dideoxy-, diacetate
PubChem CID 2734733
IUPAC Name [(2S,3S,4S)-3-acetyloxy-2-methyl-3,4-dihydro-2H-pyran-4-yl] acetate
SMILES CC1C(C(C=CO1)OC(=O)C)OC(=O)C

Specifications

Color Yellow
Quantity 5 g
Formula Weight 214.22
Percent Purity ≥95.0% (GC)
Physical Form Liquid
Chemical Name or Material 3,4-Di-O-acetyl-6-deoxy-L-glucal