missing translation for 'onlineSavingsMsg'
Learn More

3,4-Di-O-acetyl-6-deoxy-L-glucal, 98%

Catalog No. AAJ6694403
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAJ6694403 1 g
AAJ6694406 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAJ6694403 Supplier Thermo Scientific Chemicals Supplier No. J6694403

CAS: 34819-86-8 | C10H14O5 | 214.217 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 34819-86-8
Molecular Formula C10H14O5
Molecular Weight (g/mol) 214.217
MDL Number MFCD00074970
InChI Key NDEGMKQAZZBNBB-JUWDTYFHSA-N
Synonym 3,4-di-o-acetyl-6-deoxy-l-glucal, 3,4-di-o-acetyl-l-rhamnal, 2s,3s,4s-2-methyl-3,4-dihydro-2h-pyran-3,4-diyl diacetate, 3,4-di-o-acetyl-1,5-anhydro-2,6-dideoxy-l-arabino-hex-1-enitol, di-o-acetyl-l-rhamnal, 2s,3s,4s-4-acetyloxy-2-methyl-3,4-dihydro-2h-pyran-3-yl acetate, diacetyl-l-rhamnal, l-rhamnal diacetate, 6-deoxy-l-glucal diacetate, l-arabino-hex-1-enitol, 1,5-anhydro-2,6-dideoxy-, diacetate
PubChem CID 2734733
IUPAC Name [(2S,3S,4S)-3-acetyloxy-2-methyl-3,4-dihydro-2H-pyran-4-yl] acetate
SMILES CC1C(C(C=CO1)OC(=O)C)OC(=O)C

Specifications

Color Colorless to Yellow
Density 1.116
Boiling Point 68°C to 69°C
Flash Point >110°C (235°F)
Refractive Index 1.454
Quantity 1 g
Beilstein 84954
Formula Weight 214.22
Percent Purity 98%
Physical Form Viscous Liquid
Chemical Name or Material 3,4-Di-O-acetyl-6-deoxy-L-glucal
Show More Show Less
EINECSNumber 252-228-7
TSCA No
Recommended Storage Keep cold

RUO – Research Use Only