missing translation for 'onlineSavingsMsg'
Learn More

3-(4-chlorophenyl)-1H-pyrazol-5-amine, 97%, Thermo Scientific™

Catalog No. KM01548DA
Change view
Click to view available options
Quantity:
1 g
Packaging:
Amber Glass Bottle
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
KM01548DA 1 g Amber Glass Bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. KM01548DA Supplier Thermo Scientific Chemicals Supplier No. KM01548DA

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 78583-81-0
Molecular Formula C9H8ClN3
Molecular Weight (g/mol) 193.63
MDL Number MFCD00053046
InChI Key XQPBZIITFQHIDI-UHFFFAOYSA-N
Synonym 3-4-chlorophenyl-1h-pyrazol-5-amine, 5-4-chlorophenyl-1h-pyrazol-3-amine, 5-amino-3-4-chlorophenyl pyrazole, 5-4-chlorophenyl-2h-pyrazol-3-amine, 3-amino-5-4-chlorophenyl-1h-pyrazole, 5-amino-3-4-chlorophenyl-1h-pyrazole, 3-amino-5-4-chlorophenyl pyrazole, 5-4-chlorophenyl-2h-pyrazol-3-ylamine, 3-4-chlorophenyl pyrazole-5-ylamine, pubchem12126
PubChem CID 2735305
IUPAC Name 5-(4-chlorophenyl)-1H-pyrazol-3-amine
SMILES NC1=NNC(=C1)C1=CC=C(Cl)C=C1

Specifications

CAS Min % 97.0
Assay Percent Range 97%
Packaging Amber Glass Bottle
Quantity 1 g
Formula Weight 193.64
Percent Purity 97%
Chemical Name or Material 3-(4-Chlorophenyl)-1H-pyrazol-5-amine
Hazard Statement GHS H Statement:
Causes skin irritation.
May cause respiratory irritation.
Causes serious eye irritation.
Precautionary Statement GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.