missing translation for 'onlineSavingsMsg'
Learn More

3-(4-Chlorophenoxy)piperidine, 97%

Catalog No. AAH50935MD
Change view
Click to view available options
Quantity:
250 mg
1 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAH50935MD 250 mg
AAH5093503 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAH50935MD Supplier Thermo Scientific Chemicals Supplier No. H50935MD

CAS: 384346-27-4 | C11H14ClNO | 211.689 g/mol

It is employed as intermediate for pharmaceutical.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
It is employed as intermediate for pharmaceutical.

Solubility
Soluble in water.

Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Store away from oxidizing agent.

Chemical Identifiers

CAS 384346-27-4
Molecular Formula C11H14ClNO
Molecular Weight (g/mol) 211.689
MDL Number MFCD08061075
InChI Key OXELOOCWZRBKSV-UHFFFAOYSA-N
Synonym 3-4-chlorophenoxy piperidine, 3-4-chloro-phenoxy-piperidine, acmc-20ankc, piperidine,3-4-chlorophenoxy, 4-chloro-1-3-piperidyloxy benzene
PubChem CID 3865138
IUPAC Name 3-(4-chlorophenoxy)piperidine
SMILES C1CC(CNC1)OC2=CC=C(C=C2)Cl

Specifications

Quantity 250 mg
Solubility Information Soluble in water.
Formula Weight 211.69
Percent Purity 97%
Chemical Name or Material 3-(4-Chlorophenoxy)piperidine
Hazard Category H315-H319-H335
Hazard Statement GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only