missing translation for 'onlineSavingsMsg'
Learn More

3-(4-Chlorobenzoyl)propionic Acid 98.0+%, TCI America™

Catalog No. C127025G
Change view
Click to view available options
Quantity:
25 g
100 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
C127025G 25 g
C1270100G 100 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. C127025G Supplier TCI America Supplier No. C127025G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 3984-34-7
Molecular Formula C10H9ClO3
Molecular Weight (g/mol) 212.63
MDL Number MFCD00002794
InChI Key AHVASTJJVAYFPY-UHFFFAOYSA-N
Synonym 4-4-chlorophenyl-4-oxobutanoic acid, 3-4-chlorobenzoyl propionic acid, 4-4-chloro-phenyl-4-oxo-butyric acid, 3-4-chlorobenzoyl propanoic acid, 3-p-chlorobenzoyl propionic acid, 4-4-chlorophenyl-4-oxobutanoicacid, maybridge1_006214, ncistruc1_000312, ncistruc2_000239, asischem d51605
PubChem CID 77604
IUPAC Name 4-(4-chlorophenyl)-4-oxobutanoic acid
SMILES OC(=O)CCC(=O)C1=CC=C(Cl)C=C1

Specifications

Melting Point 130°C
Color White-Yellow
Quantity 25 g
Formula Weight 212.63
Percent Purity ≥98.0% (HPLC,T)
Physical Form Crystalline Powder
Chemical Name or Material 3-(4-Chlorobenzoyl)propionic Acid