missing translation for 'onlineSavingsMsg'
Learn More

3-(4-Chloro-3-fluorophenyl)propionic acid, 96%

Catalog No. p-7032654
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAH3420503 1 g
AAH3420506 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAH3420503 Supplier Thermo Scientific Chemicals Supplier No. H3420503

CAS: 881189-65-7 | C9H8ClFO2 | 202.609 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 881189-65-7
Molecular Formula C9H8ClFO2
Molecular Weight (g/mol) 202.609
MDL Number MFCD04116058
InChI Key GDXMVKNNJGWZFN-UHFFFAOYSA-N
Synonym 3-4-chloro-3-fluorophenyl propionic acid, 3-4-chloro-3-fluorophenyl propanoic acid, benzenepropanoic acid, 4-chloro-3-fluoro, 3-4-chloro-3-fluoro-phenyl-propionic acid, 4-chloro-3-fluorobenzenepropanoic acid, 3-4-chloro-3-fluorophenyl propionicacid
PubChem CID 4680108
IUPAC Name 3-(4-chloro-3-fluorophenyl)propanoic acid
SMILES C1=CC(=C(C=C1CCC(=O)O)F)Cl

Specifications

Melting Point 80°C to 84°C
Quantity 1 g
Formula Weight 202.61
Percent Purity 96%
Chemical Name or Material 3-(4-Chloro-3-fluorophenyl)propionic acid
Hazard Category H302-H315-H319-H335
Hazard Statement GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P261-P264b-P270-P271-P280-P301+P312-P302+P352-P304+P340-P305+P351+P338-P312-P330-P332+P313-P362-P501c
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only