missing translation for 'onlineSavingsMsg'
Learn More

3-(4-Bromophenyl)propionic Acid 98.0+%, TCI America™

Catalog No. B37535G
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
B37535G 5 g
B37531G 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. B37535G Supplier TCI America Supplier No. B37535G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 1643-30-7
Molecular Formula C9H9BrO2
Molecular Weight (g/mol) 229.073
MDL Number MFCD01310793
InChI Key NCSTWHYWOVZDOC-UHFFFAOYSA-N
Synonym 3-4-bromophenyl propionic acid, 3-4-bromophenyl propanoic acid, benzenepropanoic acid, 4-bromo, 3-4-bromo-phenyl-propionic acid, p-bromohydrocinnamic acid, 3-4-bromophenyl-propionic acid, 3-4-bromophenyl propanoicacid, pubchem19769, 4-bromohydrocinnamic acid, acmc-1c0vm
PubChem CID 2735609
IUPAC Name 3-(4-bromophenyl)propanoic acid
SMILES C1=CC(=CC=C1CCC(=O)O)Br

Specifications

Melting Point 134°C
Color White-Yellow
Boiling Point 250°C
Quantity 5 g
Formula Weight 229.07
Percent Purity ≥98.0% (GC,T)
Physical Form Crystalline Powder
Chemical Name or Material 3-(4-Bromophenyl)propionic Acid