missing translation for 'onlineSavingsMsg'
Learn More
Learn More
3-(4-Bromophenyl)propionic acid, 97%, Thermo Scientific™
Description
This Thermo Scientific brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.
Specifications
Specifications
| CAS Min % | 96.0 |
| CAS Max % | 100.0 |
| Infrared Spectrum | Authentic |
| Assay Percent Range | 96% min. (HPLC) |
| Packaging | Glass bottle |
| Linear Formula | BrC6H4(CH2)2COOH |
| Quantity | 1g |
| Formula Weight | 229.1 |
| Percent Purity | 97% |
| Chemical Name or Material | 3-(4-Bromophenyl)propionic acid |
Safety and Handling
| Hazard Category | GHS Signal Word: Warning |
| Hazard Statement | GHS H Statement Causes skin irritation. Causes serious eye irritation. May cause respiratory irritation. Harmful if swallowed. |
| Precautionary Statement | GHS P Statement Avoid breathing dust/fume/gas/mist/vapors/spray. IF ON SKIN: Wash with plenty of soap and water. Wear protective gloves/protective clothing/eye protection/face protection. IF IN EYES: Rinse cautiously with water for |
RUO – Research Use Only