missing translation for 'onlineSavingsMsg'
Learn More

3-(4-Bromophenyl)propionic acid, 97%, Thermo Scientific™

Catalog No. AC399450010
Change view
Click to view available options
Quantity:
1g
Packaging:
Glass bottle
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
AC399450010 1g Glass bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. AC399450010 Supplier Thermo Scientific Chemicals Supplier No. 399450010

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

missing translation for 'validMainframeMsg'

This Thermo Scientific brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

Chemical Identifiers

CAS 1643-30-7
Molecular Formula C9H9BrO2
Molecular Weight (g/mol) 229.1
MDL Number MFCD01310793
InChI Key NCSTWHYWOVZDOC-UHFFFAOYSA-N
Synonym 3-4-bromophenyl propionic acid, 3-4-bromophenyl propanoic acid, benzenepropanoic acid, 4-bromo, 3-4-bromo-phenyl-propionic acid, p-bromohydrocinnamic acid, 3-4-bromophenyl-propionic acid, 3-4-bromophenyl propanoicacid, pubchem19769, 4-bromohydrocinnamic acid, acmc-1c0vm
PubChem CID 2735609
IUPAC Name 3-(4-bromophenyl)propanoic acid
SMILES C1=CC(=CC=C1CCC(=O)O)Br

Specifications

CAS Min % 96.0
CAS Max % 100.0
Infrared Spectrum Authentic
Assay Percent Range 96% min. (HPLC)
Packaging Glass bottle
Linear Formula BrC6H4(CH2)2COOH
Quantity 1g
Formula Weight 229.1
Percent Purity 97%
Chemical Name or Material 3-(4-Bromophenyl)propionic acid
Hazard Category GHS Signal Word: Warning
Hazard Statement GHS H Statement
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Harmful if swallowed.
Precautionary Statement GHS P Statement
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for

RUO – Research Use Only