missing translation for 'onlineSavingsMsg'
Learn More
Learn More
3-(4-Bromophenyl)-9-phenylcarbazole 98.0+%, TCI America™
$54.06 - $178.95
Chemical Identifiers
| CAS | 1028647-93-9 |
|---|---|
| Molecular Formula | C24H16BrN |
| Molecular Weight (g/mol) | 398.303 |
| MDL Number | MFCD14582939 |
| InChI Key | JEYLGFCAZBGCMC-UHFFFAOYSA-N |
| Synonym | 3-4-bromophenyl-9-phenyl-9h-carbazole, 3-4-bromophenyl-9-phenylcarbazole, 3-4-bromophenyl-n-phenylcarbazole, 3-4-bromophenyl-9-phenyl9h-carbazole, pubchem23233, 3-4-bromophenyl-9-phenyl-carbazole, 3-4-bromo-phenyl-9-phenyl-9h-carbazole |
| PubChem CID | 51358293 |
| IUPAC Name | 3-(4-bromophenyl)-9-phenylcarbazole |
| SMILES | C1=CC=C(C=C1)N2C3=C(C=C(C=C3)C4=CC=C(C=C4)Br)C5=CC=CC=C52 |
Chemical Identifiers
| 1028647-93-9 | |
| 398.303 | |
| JEYLGFCAZBGCMC-UHFFFAOYSA-N | |
| 51358293 | |
| C1=CC=C(C=C1)N2C3=C(C=C(C=C3)C4=CC=C(C=C4)Br)C5=CC=CC=C52 |
| C24H16BrN | |
| MFCD14582939 | |
| 3-4-bromophenyl-9-phenyl-9h-carbazole, 3-4-bromophenyl-9-phenylcarbazole, 3-4-bromophenyl-n-phenylcarbazole, 3-4-bromophenyl-9-phenyl9h-carbazole, pubchem23233, 3-4-bromophenyl-9-phenyl-carbazole, 3-4-bromo-phenyl-9-phenyl-9h-carbazole | |
| 3-(4-bromophenyl)-9-phenylcarbazole |
Specifications
| 1028647-93-9 | |
| White-Yellow | |
| MFCD14582939 | |
| 3-4-bromophenyl-9-phenyl-9h-carbazole, 3-4-bromophenyl-9-phenylcarbazole, 3-4-bromophenyl-n-phenylcarbazole, 3-4-bromophenyl-9-phenyl9h-carbazole, pubchem23233, 3-4-bromophenyl-9-phenyl-carbazole, 3-4-bromo-phenyl-9-phenyl-9h-carbazole | |
| C1=CC=C(C=C1)N2C3=C(C=C(C=C3)C4=CC=C(C=C4)Br)C5=CC=CC=C52 | |
| 398.303 | |
| 398.30 | |
| Crystalline Powder |
| 138°C | |
| C24H16BrN | |
| 1 g | |
| JEYLGFCAZBGCMC-UHFFFAOYSA-N | |
| 3-(4-bromophenyl)-9-phenylcarbazole | |
| 51358293 | |
| ≥98.0% (GC) | |
| 3-(4-Bromophenyl)-9-phenylcarbazole |
Safety and Handling
TSCA : No