missing translation for 'onlineSavingsMsg'
Learn More

3-(4-Bromophenyl)-9-phenylcarbazole 98.0+%, TCI America™

Catalog No. B44525G
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
B44525G 5 g
B44521G 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. B44525G Supplier TCI America Supplier No. B44525G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 1028647-93-9
Molecular Formula C24H16BrN
Molecular Weight (g/mol) 398.303
MDL Number MFCD14582939
InChI Key JEYLGFCAZBGCMC-UHFFFAOYSA-N
Synonym 3-4-bromophenyl-9-phenyl-9h-carbazole, 3-4-bromophenyl-9-phenylcarbazole, 3-4-bromophenyl-n-phenylcarbazole, 3-4-bromophenyl-9-phenyl9h-carbazole, pubchem23233, 3-4-bromophenyl-9-phenyl-carbazole, 3-4-bromo-phenyl-9-phenyl-9h-carbazole
PubChem CID 51358293
IUPAC Name 3-(4-bromophenyl)-9-phenylcarbazole
SMILES C1=CC=C(C=C1)N2C3=C(C=C(C=C3)C4=CC=C(C=C4)Br)C5=CC=CC=C52

Specifications

Melting Point 138°C
Color White-Yellow
Quantity 5 g
Formula Weight 398.30
Percent Purity ≥98.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material 3-(4-Bromophenyl)-9-phenylcarbazole