missing translation for 'onlineSavingsMsg'
Learn More

3-[4-(Bromomethyl)phenyl]-5-methyl-1,2,4-oxadiazole 97+%, Thermo Scientific™

Catalog No. CC34808CB
Change view
Click to view available options
Quantity:
250mg
1g
Packaging:
Amber Glass Bottle
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
CC34808CB 250mg Amber Glass Bottle
CC34808DA 1g Amber Glass Bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. CC34808CB Supplier Thermo Scientific Chemicals Supplier No. CC34808CB

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 256956-42-0
Molecular Formula C10H9BrN2O
Molecular Weight (g/mol) 253.099
InChI Key BMXSEDLZMUPKSH-UHFFFAOYSA-N
Synonym 3-4-bromomethyl phenyl-5-methyl-1,2,4-oxadiazole, 1,2,4-oxadiazole, 3-4-bromomethyl phenyl-5-methyl, 1,2,4-oxadiazole,3-4-bromomethyl phenyl-5-methyl, 3-4-bromomethyl phenyl-5-methyl-1,2,4-oxadiazol
PubChem CID 7164590
IUPAC Name 3-[4-(bromomethyl)phenyl]-5-methyl-1,2,4-oxadiazole
SMILES CC1=NC(=NO1)C2=CC=C(C=C2)CBr

Specifications

Assay Percent Range 97%
Packaging Amber Glass Bottle
Quantity 250mg
Formula Weight 253.1
Percent Purity 97%
Chemical Name or Material 3-[4-(Bromomethyl)phenyl]-5-methyl-1, 2, 4-oxadiazole