missing translation for 'onlineSavingsMsg'
Learn More

3,4-Bis(trifluoromethyl)pyridine, 96%, Thermo Scientific Chemicals

Catalog No. p-7028615
Change view
Click to view available options
Quantity:
1 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAH2600803 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. AAH2600803 Supplier Thermo Scientific Chemicals Supplier No. H2600803
Only null left

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 20857-46-9
Molecular Formula C7H3F6N
Molecular Weight (g/mol) 215.10
MDL Number MFCD08686956
InChI Key SVYGFFYEEPFDOC-UHFFFAOYSA-N
Synonym 3,4-bis trifluoromethyl pyridine, acmc-20anfx, 3,4-di trifluoromethyl pyridine, pyridine,3,4-bis trifluoromethyl
PubChem CID 14937917
IUPAC Name 3,4-bis(trifluoromethyl)pyridine
SMILES FC(F)(F)C1=C(C=NC=C1)C(F)(F)F

Specifications

Boiling Point 42°C to 44°C (24 mmHg)
Refractive Index 1.392
Quantity 1 g
Formula Weight 215.1
Percent Purity 96%
Chemical Name or Material 3,4-Bis(trifluoromethyl)pyridine
Hazard Category H315-H319-H335
Hazard Statement GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only