missing translation for 'onlineSavingsMsg'
Learn More

3-(4-Biphenylyl)-N-Fmoc-L-alanine, 95%

Catalog No. AAH52002MD
Change view
Click to view available options
Quantity:
250 mg
1 g
5 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAH52002MD 250 mg
AAH5200203 1 g
AAH5200206 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AAH52002MD Supplier Thermo Scientific Chemicals Supplier No. H52002MD
Get Up to $1,800 in Chemicals with Purchase
View all promotions

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 199110-64-0 | C30H25NO4 | 463.533 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 199110-64-0
Molecular Formula C30H25NO4
Molecular Weight (g/mol) 463.533
MDL Number MFCD00191198
InChI Key VSGACONKQRJFGX-NDEPHWFRSA-N
Synonym fmoc-l-4,4'-biphenylalanine, s-2-9h-fluoren-9-yl methoxy carbonyl amino-3-1,1'-biphenyl-4-yl propanoic acid, fmoc-bip 4,4'-oh, fmoc-4-biphenyl-l-ala, fmoc-4-phenyl-phe-oh, 3-4-biphenylyl-n-fmoc-l-alanine, fmoc-p-phenyl-l-phenylalanine, fmoc-bip-oh, fmoc-l-phe 4-ph-oh, 2s-3-1,1'-biphenyl-4-yl-2-9h-fluoren-9-ylmethoxy carbonyl amino propanoic acid
PubChem CID 2761797
IUPAC Name (2S)-2-(9H-fluoren-9-ylmethoxycarbonylamino)-3-(4-phenylphenyl)propanoic acid
SMILES C1=CC=C(C=C1)C2=CC=C(C=C2)CC(C(=O)O)NC(=O)OCC3C4=CC=CC=C4C5=CC=CC=C35

Specifications

Quantity 250 mg
Formula Weight 463.52
Percent Purity 95%
Chemical Name or Material 3-(4-Biphenylyl)-N-Fmoc-L-alanine
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only