missing translation for 'onlineSavingsMsg'
Learn More

3-(4-Biphenyl)propionic acid, 98%

Catalog No. AAH3272903
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAH3272903 1 g
AAH3272906 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAH3272903 Supplier Thermo Scientific Chemicals Supplier No. H3272903
Get Up to $1,800 in Chemicals with Purchase
View all promotions

CAS: 35888-99-4 | C15H14O2 | 226.28 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 35888-99-4
Molecular Formula C15H14O2
Molecular Weight (g/mol) 226.28
MDL Number MFCD06823979
InChI Key MVFHRQWYCXYYMU-UHFFFAOYSA-N
Synonym 2 4-biphenyl propionic acid, 3-4-phenylphenyl propanoic acid, 3-4-biphenyl propionic acid, 3-1,1'-biphenyl-4-yl propanoic acid, 3-biphenyl-4-yl propanoic acid, 1,1'-biphenyl-4-propanoic acid, 2 para-biphenyl propionic acid, biphenyl-4-propanoic acid
PubChem CID 3082434
IUPAC Name 3-(4-phenylphenyl)propanoic acid
SMILES OC(=O)CCC1=CC=C(C=C1)C1=CC=CC=C1

Specifications

Melting Point 149°C to 152°C
Quantity 1 g
Formula Weight 226.28
Percent Purity 98%
Chemical Name or Material 3-(4-Biphenyl)propionic acid
Hazard Category H315-H319-H335
Hazard Statement GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only