missing translation for 'onlineSavingsMsg'
Learn More

3,4,9,10-Perylenetetracarboxylic diimide

Catalog No. p-7059159
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AA4409806 5 g
AA4409814 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AA4409806 Supplier Thermo Scientific Chemicals Supplier No. 04409806

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 81-33-4 | C24H10N2O4 | 390.354 g/mol

Used in chemical research. PTCDI is used extensively as an industrial pigment. It is used as tunable laser dye, light-harvesting material, transistor, molecular switched, solar cell, and optoelectronic device. Planar perylene- and naphthalene-based diimide linkers can be employed to tether the Watson- Crick and the Hoogsteen strands of a DNA triplex, thus providing conjugates capable of targeting singlestranded nucleic acids with the formation of hairpin triplexes.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 81-33-4
Molecular Formula C24H10N2O4
Molecular Weight (g/mol) 390.354
MDL Number MFCD00024144
InChI Key KJOLVZJFMDVPGB-UHFFFAOYSA-N
Synonym perylimid, 3,4,9,10-perylenetetracarboxylic diimide, perylenediimide, 3,4,9,10-perylenetetracarboxylic acid diimide, perylene-3,4:9,10-tetracarboxydiimide, unii-63nli8842l, anthra 2,1,9-def:6,5,10-d'e'f' diisoquinoline-1,3,8,10 2h,9h-tetrone, anthra 2,1,9-def:6,5,10-d'e'f' diisoquinoline-1,3,8,10 2h,9h-tetraone, isoquino 4',5',6':6,5,10 anthra 2,1,9-def isoquinoline-1,3,8,10 2h,9h-tetrone
PubChem CID 66475
ChEBI CHEBI:52753
SMILES C1=CC2=C3C(=CC=C4C3=C1C5=C6C4=CC=C7C6=C(C=C5)C(=O)NC7=O)C(=O)NC2=O

Specifications

Quantity 5 g
Solubility Information Insoluble in water.
Formula Weight 390.35
Physical Form Powder
Chemical Name or Material 3,4,9,10-Perylenetetracarboxylic diimide
EINECSNumber 201-344-6
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only