missing translation for 'onlineSavingsMsg'
Learn More

3,4,9,10-Perylenetetracarboxylic Diimide 95.0+%, TCI America™

Catalog No. P098425G
Change view
Click to view available options
Quantity:
25 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
P098425G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. P098425G Supplier TCI America Supplier No. P098425G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 81-33-4
Molecular Formula C24H10N2O4
Molecular Weight (g/mol) 390.354
MDL Number MFCD00024144
InChI Key KJOLVZJFMDVPGB-UHFFFAOYSA-N
Synonym perylimid, 3,4,9,10-perylenetetracarboxylic diimide, perylenediimide, 3,4,9,10-perylenetetracarboxylic acid diimide, perylene-3,4:9,10-tetracarboxydiimide, unii-63nli8842l, anthra 2,1,9-def:6,5,10-d'e'f' diisoquinoline-1,3,8,10 2h,9h-tetrone, anthra 2,1,9-def:6,5,10-d'e'f' diisoquinoline-1,3,8,10 2h,9h-tetraone, isoquino 4',5',6':6,5,10 anthra 2,1,9-def isoquinoline-1,3,8,10 2h,9h-tetrone
PubChem CID 66475
ChEBI CHEBI:52753
SMILES C1=CC2=C3C(=CC=C4C3=C1C5=C6C4=CC=C7C6=C(C=C5)C(=O)NC7=O)C(=O)NC2=O

Specifications

Quantity 25 g
Formula Weight 390.35
Percent Purity ≥95.0% (N)
Physical Form Crystalline Powder
Chemical Name or Material 3,4,9,10-Perylenetetracarboxylic Diimide
EINECSNumber (5)-1111&(5)-3326&(5)-5211
TSCA Yes