Learn More
3,4,9,10-Perylenetetracarboxylic dianhydride, 98%
CAS: 128-69-8 | C24H8O6 | 392.32 g/mol
Supplier: Thermo Scientific Chemicals L1437018
Description
3,4,9,10-Perylenetetracarboxylic dianhydride is an important raw material and intermediate used in organic synthesis, pharmaceuticals and agrochemicals. It is mainly used in air dried and baking enamels, epoxy, polyurethane and polyester based paints. It is also used in printing inks for bank and security documents.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications3,4,9,10-Perylenetetracarboxylic dianhydride is an important raw material and intermediate used in organic synthesis, pharmaceuticals and agrochemicals. It is mainly used in air dried and baking enamels, epoxy, polyurethane and polyester based paints. It is also used in printing inks for bank and security documents.
Solubility
Soluble in water (0.87g/L at 20°C).
Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. It is sensitive to moisture. It is hygroscopic in nature.
Specifications
| 3,4,9,10-Perylenetetracarboxylic dianhydride | |
| >300°C | |
| C24H8O6 | |
| 50 g | |
| Moisture sensitive | |
| Soluble in water (0.87g/L at 20°C). | |
| O=C1OC(=O)C2=CC=C3C4=CC=C5C(=O)OC(=O)C6=CC=C(C7=CC=C1C2=C37)C4=C56 | |
| 392.32 | |
| 392.32 |
| 128-69-8 | |
| Odorless | |
| MFCD00006916 | |
| 57831 | |
| 3,4,9,10-perylenetetracarboxylic dianhydride, pigment red 224, ptcda, perylene-3,4,9,10-tetracarboxylic dianhydride, perylenetetracarboxylic anhydride, anthra 2,1,9-def:6,5,10-d'e'f' diisochromene-1,3,8,10-tetraone, perylo 3,4-cd:9,10-c'd' dipyran-1,3,8,10-tetrone, perylenetetracarboxylic acid dianhydride, 3,4:9,10-perylenetetracarboxylic anhydride | |
| CLYVDMAATCIVBF-UHFFFAOYSA-N | |
| 7,18-dioxaheptacyclo[14.6.2.2²,âµ.0³,¹².0â´,â¹.0¹³,²³.0²â°,²â´]hexacosa-1(22),2(26),3,5(25),9,11,13,15,20,23-decaene-6,8,17,19-tetrone | |
| 67191 | |
| 98% |
Chemical Identifiers
| 128-69-8 | |
| 392.32 | |
| CLYVDMAATCIVBF-UHFFFAOYSA-N | |
| 67191 | |
| O=C1OC(=O)C2=CC=C3C4=CC=C5C(=O)OC(=O)C6=CC=C(C7=CC=C1C2=C37)C4=C56 |
| C24H8O6 | |
| MFCD00006916 | |
| 3,4,9,10-perylenetetracarboxylic dianhydride, pigment red 224, ptcda, perylene-3,4,9,10-tetracarboxylic dianhydride, perylenetetracarboxylic anhydride, anthra 2,1,9-def:6,5,10-d'e'f' diisochromene-1,3,8,10-tetraone, perylo 3,4-cd:9,10-c'd' dipyran-1,3,8,10-tetrone, perylenetetracarboxylic acid dianhydride, 3,4:9,10-perylenetetracarboxylic anhydride | |
| 7,18-dioxaheptacyclo[14.6.2.2²,âµ.0³,¹².0â´,â¹.0¹³,²³.0²â°,²â´]hexacosa-1(22),2(26),3,5(25),9,11,13,15,20,23-decaene-6,8,17,19-tetrone |
Safety and Handling
GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.
Warning
EINECSNumber : 204-905-3
TSCA : Yes
Recommended Storage : Ambient temperatures; Store under Nitrogen
RUO – Research Use Only