missing translation for 'onlineSavingsMsg'
Learn More

3,4,9,10-Perylenetetracarboxylic dianhydride, 98%

Catalog No. AAL1437009
Change view
Click to view available options
Quantity:
10 g
50 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAL1437009 10 g
AAL1437018 50 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAL1437009 Supplier Thermo Scientific Chemicals Supplier No. L1437009
Get Up to $1,800 in Chemicals with Purchase
View all promotions

CAS: 128-69-8 | C24H8O6 | 392.32 g/mol

3,4,9,10-Perylenetetracarboxylic dianhydride is an important raw material and intermediate used in organic synthesis, pharmaceuticals and agrochemicals. It is mainly used in air dried and baking enamels, epoxy, polyurethane and polyester based paints. It is also used in printing inks for bank and security documents.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
3,4,9,10-Perylenetetracarboxylic dianhydride is an important raw material and intermediate used in organic synthesis, pharmaceuticals and agrochemicals. It is mainly used in air dried and baking enamels, epoxy, polyurethane and polyester based paints. It is also used in printing inks for bank and security documents.

Solubility
Soluble in water (0.87g/L at 20°C).

Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. It is sensitive to moisture. It is hygroscopic in nature.

Chemical Identifiers

CAS 128-69-8
Molecular Formula C24H8O6
Molecular Weight (g/mol) 392.32
MDL Number MFCD00006916
InChI Key CLYVDMAATCIVBF-UHFFFAOYSA-N
Synonym 3,4,9,10-perylenetetracarboxylic dianhydride, pigment red 224, ptcda, perylene-3,4,9,10-tetracarboxylic dianhydride, perylenetetracarboxylic anhydride, anthra 2,1,9-def:6,5,10-d'e'f' diisochromene-1,3,8,10-tetraone, perylo 3,4-cd:9,10-c'd' dipyran-1,3,8,10-tetrone, perylenetetracarboxylic acid dianhydride, 3,4:9,10-perylenetetracarboxylic anhydride
PubChem CID 67191
IUPAC Name 7,18-dioxaheptacyclo[14.6.2.2²,⁵.0³,¹².0⁴,⁹.0¹³,²³.0²⁰,²⁴]hexacosa-1(22),2(26),3,5(25),9,11,13,15,20,23-decaene-6,8,17,19-tetrone
SMILES O=C1OC(=O)C2=CC=C3C4=CC=C5C(=O)OC(=O)C6=CC=C(C7=CC=C1C2=C37)C4=C56

Specifications

Melting Point >300°C
Odor Odorless
Quantity 10 g
Beilstein 57831
Sensitivity Moisture sensitive
Solubility Information Soluble in water (0.87g/L at 20°C).
Formula Weight 392.32
Percent Purity 98%
Chemical Name or Material 3,4,9,10-Perylenetetracarboxylic dianhydride
Hazard Category Warning
Hazard Statement GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.
EINECSNumber 204-905-3
TSCA Yes
Recommended Storage Ambient temperatures; Store under Nitrogen

RUO – Research Use Only