missing translation for 'onlineSavingsMsg'
Learn More

3,4,6-Tri-O-methyl-D-glucal, 97%

Catalog No. AAH60694MD
Change view
Click to view available options
Quantity:
250 mg
1 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAH60694MD 250 mg
AAH6069403 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAH60694MD Supplier Thermo Scientific Chemicals Supplier No. H60694MD
Only null left

CAS: 16740-98-0 | C9H16O4 | 188.22 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 16740-98-0
Molecular Formula C9H16O4
Molecular Weight (g/mol) 188.22
MDL Number MFCD22989010
InChI Key PZJPNKKRMNFDQJ-UHFFFAOYNA-N
Synonym tri-o-methyl-d-glucal, 3,4,6-tri-o-methyl-d-glucal, tri-o-methyl glucal, 2r,3s,4r-3,4-dimethoxy-2-methoxymethyl-3,4-dihydro-2h-pyran, 3-o,4-o,6-o-trimethyl-1,2-dideoxy-d-arabino-1-hexenopyranose
PubChem CID 11252452
SMILES COCC1OC=CC(OC)C1OC

Specifications

Quantity 250 mg
IUPAC Name 3,4-dimethoxy-2-(methoxymethyl)-3,4-dihydro-2H-pyran
Formula Weight 188.22
Percent Purity 97%
Chemical Name or Material 3,4,6-Tri-O-methyl-D-glucal
TSCA No
Recommended Storage Keep cold

RUO – Research Use Only