missing translation for 'onlineSavingsMsg'
Learn More

3,4,6-Tri-O-acetyl-D-galactal

Catalog No. p-7043288
Change view
Click to view available options
Quantity:
1 g
5 g
10 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAJ6690403 1 g
AAJ6690406 5 g
AAJ6690409 10 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AAJ6690403 Supplier Thermo Scientific Chemicals Supplier No. J6690403
Only null left

CAS: 4098-06-0 | C12H16O7 | 272.25 g/mol

3,4,6-Tri-O-acetyl-D-galactal is important building block for both solution- and solid-phase synthesis of oligosaccharides.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
3,4,6-Tri-O-acetyl-D-galactal is important building block for both solution- and solid-phase synthesis of oligosaccharides.

Solubility
Soluble in chloroform at 10mg/ml

Notes
Keep container tightly closed. Store in cool, dry conditions in well sealed containers.

Chemical Identifiers

CAS 4098-06-0
Molecular Formula C12H16O7
Molecular Weight (g/mol) 272.25
MDL Number MFCD00064092
InChI Key LLPWGHLVUPBSLP-UHFFFAOYNA-N
Synonym 3,4,6-tri-o-acetyl-d-galactal, tri-o-acetyl-d-galactal, 2r,3r,4r-2-acetoxymethyl-3,4-dihydro-2h-pyran-3,4-diyl diacetate, d-arabino-hex-5-enitol, 2,6-anhydro-5-deoxy-, triacetate, 1,3,4-tri-o-acetyl-2,6-anhydro-5-deoxyhex-5-enitol, d-galactal triacetate, 2,6-anhydro-5-deoxy-d-arabino-hex-5-enitol triacetate, 2r,3r,4r-3,4-bis acetyloxy-3,4-dihydro-2h-pyran-2-yl methyl acetate, galactal triacetate, tri-o-acetylgalactal
PubChem CID 640125
IUPAC Name [(2R,3R,4R)-3,4-diacetyloxy-3,4-dihydro-2H-pyran-2-yl]methyl acetate
SMILES CC(=O)OCC1OC=CC(OC(C)=O)C1OC(C)=O

Specifications

Melting Point 34°C to 38°C
Color Yellow
Flash Point >110°C (235°F)
Quantity 1 g
Solubility Information Soluble in chloroform at 10mg/ml
Formula Weight 272.25
Physical Form Oil or Solid
Chemical Name or Material 3,4,6-Tri-O-acetyl-D-galactal
EINECSNumber 223-859-5
TSCA No
Recommended Storage Keep cold

RUO – Research Use Only