missing translation for 'onlineSavingsMsg'
Learn More

3,4,5-Tris(dodecyloxy)benzaldehyde 98.0+%, TCI America™

Catalog No. p-7137819
Change view
Click to view available options
Quantity:
200 mg
1 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
T3624200MG 200 mg
T36241G 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. T3624200MG Supplier TCI America Supplier No. T3624200MG
Only null left

Chemical Identifiers

CAS 117241-32-4
Molecular Formula C43H78O4
Molecular Weight (g/mol) 659.093
MDL Number MFCD29917550
InChI Key ABDPYSIFPMBTOS-UHFFFAOYSA-N
PubChem CID 14196731
IUPAC Name 3,4,5-tridodecoxybenzaldehyde
SMILES CCCCCCCCCCCCOC1=CC(=CC(=C1OCCCCCCCCCCCC)OCCCCCCCCCCCC)C=O

Specifications

Melting Point 50°C
Quantity 200 mg
Formula Weight 659.09
Percent Purity ≥98.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material 3,4,5-Tris(dodecyloxy)benzaldehyde