missing translation for 'onlineSavingsMsg'
Learn More

3,4,5-Trimethoxybenzoic Acid 98.0+%, TCI America™

Catalog No. T070525G
Change view
Click to view available options
Quantity:
25 g
500 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
T070525G 25 g
T0705500G 500 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. T070525G Supplier TCI America Supplier No. T070525G
Only null left

Chemical Identifiers

CAS 118-41-2
Molecular Formula C10H12O5
Molecular Weight (g/mol) 212.2
MDL Number MFCD00002501
InChI Key SJSOFNCYXJUNBT-UHFFFAOYSA-N
Synonym eudesmic acid, gallic acid trimethyl ether, tri-o-methylgallic acid, trimethylgallic acid, benzoic acid, 3,4,5-trimethoxy, veratric acid, 5-methoxy, unii-v5c9h0sc9f, 3,4,5-trimethoxy-benzoic acid, 5-methoxy-veratric acid, v5c9h0sc9f
PubChem CID 8357
ChEBI CHEBI:454991
IUPAC Name 3,4,5-trimethoxybenzoic acid
SMILES COC1=CC(=CC(OC)=C1OC)C(O)=O

Specifications

Melting Point 170°C
Color White to Yellow
Boiling Point 227°C
Quantity 25 g
Formula Weight 212.2
Percent Purity ≥98.0% (GC,T)
Physical Form Crystalline Powder
Chemical Name or Material 3,4,5-Trimethoxybenzoic Acid
EINECSNumber (3)-1627
TSCA Yes